Nα,Nα-Bis(carboxymethyl)-L-lysine hydrate ≥97.0% (by TLC), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:941689-36-7
:MFCD24387945
Formula:C₁₀H₂₀N₂O₇
:280.27 g/mol
Specification Test Results
- APPEARANCE (COLOR):White
- APPEARANCE (FORM):Powder
- PURITY (TLC AREA %):≥ 97.0 %
- WATER:≤ 12 %
- PROTON NMR SPECTRUM:CONFORMS TO STRUCTURE
- REMARKS:may contain up ~2 mol water and ~10% inorganic salts
Specification Test Results
Frequently Bought Together