Benzyl viologen dichloride 97% (by HPLC), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1102-19-8
:
Formula:C₂₄H₂₂Cl₂N₂
MDL Number:MFCD00011991
:409.35 g/mol
Melting Pt:260 °C (dec., lit.)
:GHS07
Specification Test Results
- Appearance (Colour):White to Off-White
- Appearance (Form) (Powder, Crystals, Crystalline Powder and/or Chunks):Conforms to Requirements
- Infrared spectrum:Conforms to Structure
- Proton NMR spectrum:Conforms to Structure
- Titration by AgNO₃:96.5 - 103.5 %
- Purity (HPLC):≥96.5%
- Solubility (Turbidity), c = 50 mg/ml, H₂O:Clear to Very Slightly Hazy
- Solubility (Color):Colorless to Yellow
Specification Test Results
Frequently Bought Together