Order Entry
United Kingdom
ContactUsLinkComponent
Counting chambers, Neubauer
Counting chambers, Neubauer
Catalog # HAWKAS2000
Supplier:  HAWKSLEY AND SONS
Counting chambers, Neubauer
Catalog # HAWKAS2000
Supplier:  HAWKSLEY AND SONS
Specifications
  • Depth of chamber:
    0,1 mm
  • Description:
    Cell counter, 2-cell standard
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
These cell counting chambers are moulded from a single piece of thermal and shock-resistant glass.
  • 2-cell
  • Optically polished platform
An H-shaped moat forms either one or two counting areas, or plateaus. Each plateau features an etched grid or ruling. Contact of the flat, polished cover glass surfaces with the cover glass supports produces an exact volume of fluid over the counting area.
The ruled surface is 0,1 mm below the cover glass, limiting the Volume of 1 Square mm of blood or fluid over a square mm at 0,1 cu.mm.
Certifications: BS 748