Microscope stands, Dino-Lite, RK-06A / RK-06-AE
Supplier: Dino-Lite / IDCP B.V.
Certificates
About this item
These mid-range tabletop stands with square metal base are either designed for general purposes or for ESD applications. They can be used with all handheld Dino-Lite microscopes.
- Constructed of stainless steel and lightweight aluminium
- Adjustable horizontal arm has a 15 cm range with 360-degree rotation
- Smooth focus adjustment and quick vertical movement: Ease the task of focusing when using Dino-Lite microscopes
- Ease-of-assembly and adaptable extension arm design adds further mobility and flexibility to user’s applications
The RK-06-AE comes with an ESD (electrostatic discharge) safe base plate instead of the standard RK-06A base plate.
For users that require additional working distance, the optional RK-06-PL extension pole is available. It is required when working with the Dino-Lite Long Working Distance and Extra Long Working Distance microscopes.
Delivery: Supplied with additional horizontal arm RK-10-EX.
Extensions
Product Information
Frequently Bought Together