Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bottles
Glass Bottles
VWR®, Bottle, Wide Neck, Amber
VWR®, Bottle, Wide Neck, Amber
VWRLSTL8
:
LAB3 OWN PRODUCTION
VWR®, Bottle, Wide Neck, Amber
VWRLSTL8
:
LAB3 OWN PRODUCTION
Type:
Wide mouth, PTFE lined cap
Capacity:
2500 ml
Colour:
Amber
Pk:
2,5 L