Order Entry
United Kingdom
ContactUsLinkComponent
Disinfectant powder, Virkon S
Disinfectant powder, Virkon S
Catalog # DIUKANT0190
Supplier:  DISTRIBUTOR
Disinfectant powder, Virkon S
Catalog # DIUKANT0190
Supplier:  DISTRIBUTOR
Specifications
  • Description:
    Virkon S disinfectant powder
  • Seq:
    01
  • Packed:
    1 kg
  • Pk:
    1 kg
Specifications
About this item
A broad-spectrum disinfectant effective against all 18 virus families affecting man and animals, including HIV and hepatitis B.
  • Bactericide and fungicide
  • Odourless and non-irritant to skin and eyes
  • Dissolves easily
  • Non-corrosive, non-bleaching
Caution: Use biocides safely. Always read the label and product information before use.