Calbiochem® Bovine hyaluronic acid binding protein (from Nasal cartilage), Biotin
385911-50
:
- Pk:50 µG
- Conjugation:Biotin
- Source:Nasal cartilage
- Species:Bovine
- Protein/peptide name:Hyaluronic acid binding protein
Affinity purified proteins. Binds specifically and strongly to hyaluronic acid. Suitable for ELISA and histochemistry.
- Lyophilised form
- Store at –20 °C
Composed of two binding polypeptides, derived from N-terminal regions of the hyaluronic acid binding proteoglycan and linkage proteins.