Chemical resistant gloves, Camatex 451
Camatex 451 gloves are made of comfortable, highly elastic natural latex with a cotton liner.
- Excellent grip on wet surfaces
- Remain comfortable in higher or lower temperatures, even when worn for extended periods
- Fully coated - liquid proof
- Excellent mechanical resistance
- Roughened palm
Used in the food industry, construction industry, for harvesting, cleaning and steam blasting works, canteen kitchens, erection of scaffolding and in the beverage industry.
: EN 388:2016 (2131X), EN ISO 374-1 Type A (KLMPST); Cat. III; AQL 1,5
Food contact approved.
Food contact approved.
- Catalog No:
- KCHL451.7
- KCHL451.10
- KCHL451.9
- KCHL451.8
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Roughened palm
- Roughened palm
- Roughened palm
- Roughened palm
- EN norm:
- EN 388:2016 (2131X), EN ISO 374-1 Type A (KLMPST)
- EN 388:2016 (2131X), EN ISO 374-1 Type A (KLMPST)
- EN 388:2016 (2131X), EN ISO 374-1 Type A (KLMPST)
- EN 388:2016 (2131X), EN ISO 374-1 Type A (KLMPST)
- Food contact approved:
- +
- +
- +
- +
Frequently Bought Together