Nitrosamine calibration mix, method 521, (7 components)
Catalog # REST31898
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each in methylene chloride
- Pk:1 Ampoul
- Description:Nitrosamine calibrationm mix
- Pk Info:1 ml/ampoule
Specifications
About this item
This contains N-nitrosodiethylamine, N-nitrosodimethylamine, N-nitrosodi-n-butylamine, N-nitrosodi-n-propylamine, N-nitrosomethylethylamine, N-nitrosopiperidine and N-nitrosopyrrolidine.