CLP/SVOA additions mix (3 components)
Catalog # REST31902
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml in methylene chloride
- Pk:1 Ampoul
- Description:CLP/SVOA additions mix
- Pk Info:1 ml/ampoule
Specifications
About this item
This contains atrazine, benzaldehyde and ε-caprolactam.