TO-15 subset 25 component mix
Catalog # REST34434
Supplier: Restek
CAS Number:
Specifications
- Concentration:1 ppm in nitrogen
- Pk:1
- Description:TO-15 subset 25 component mix
- Pk Info:1 ml/ampoule
- Accuracy:Analytical accuracy: ±5 %
Specifications
About this item
Environmental air sampling gas standards meet lab requirements for two separate sources of calibration standards.
- High-quality
- No rental or demurrage fees required
- Fast delivery provides freshest standard possible
- One year shelf life
- Blend tolerance: ±10%
TO-15 Subset 25 component mix contains the following components: acetone; allyl chloride; benzyl chloride; bromodichloromethane; bromoform; 1,3-butadiene; 2-butanone (MEK); carbon disulfide; cyclohexane; dibromochloromethane; trans-1,2-dichloroethene; 1,4-dioxane; ethyl acetate; 4-ethyltoluene; heptane; hexane; 2-hexanone (MBK); 4-methyl-2-pentanone; methyl tert-butyl ether (MTBE); 2-propanol; propylene; tetrahydrofuran; 2,2,4-trimethylpentane; vinyl acetate; vinyl bromide.
W×H: 83×295 mm.
Weight: 0,7 kg.
Pressure: 240 psig (17 bar).
Caution: *Stability of this compound cannot be guaranteed.