Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Kovats test mix #1 (8 components)

Kovats test mix #1 (8 components)

Kovats test mix #1, 250 µg/ml each in methylene chloride, 1 ml/ampoule, 1 mL
Catalog # REST35269
Supplier:   Restek
Kovats test mix #1 (8 components)
Catalog # REST35269
Supplier:  Restek
CAS Number:  
Kovats test mix #1 (8 components)
Kovats test mix #1 (8 components)

Kovats test mix #1, 250 µg/ml each in methylene chloride, 1 ml/ampoule, 1 mL

CAS Number:  
Price
£75.30
Log in for availability
1 * 1 mL
Product Details & Documents

Contains: n-hexane, C6, n-heptane, C7, n-octane, C8, n-nonane, C9, benzene, 1-butanol, 1-nitropropane, 2-pentanone.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Specifications

Specifications

  • Concentration:
    250 µg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    Kovats test mix #1
  • Pk Info:
    1 ml/ampoule

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.