α-terpineol
Catalog # REST33912
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in methylene chloride
- Pk:1 Ampoul
- CAS No.:98-55-5
- Description:α-terpineol
- Environmentally Preferable:
- Formula:C₁₀H₁₈O
- Description:α-Terpineol
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.