USP <467> calibration mix #7
USP <467> calibration mixture #7, In dimethyl sulphoxide, 1 ml/ampoule, 1
Catalog # REST36009
Supplier: Restek
Jump to:
Product Details & Documents
The mix consists of chloroform (60 μg/ml), 1,4-dioxane (380 μg/ml), methylene chloride (dichloromethane) (600 μg/ml) and trichloroethene (80 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Specifications
Specifications
- Concentration:In dimethyl sulphoxide
- Pk:1
- Description:USP <467> calibration mixture #7
- Pk Info:1 ml/ampoule