1,1,2,2-Tetrachloroethane
Catalog # REST30412
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 μg/ml in P and T methanol
- Pk:1
- CAS No.:79-34-5
- Description:1,1,2,2-Tetrachloroethane
- Environmentally Preferable:
- Formula:C₂H₂Cl₄
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.