Order Entry
United Kingdom
ContactUsLinkComponent
ASTM® D3606 calibration standard #1 with sec-butanol internal standard (3 components)
ASTM® D3606 calibration standard #1 with sec-butanol internal standard (3 components)
Catalog # REST30661
Supplier:  Restek
CAS Number:  
undefined
ASTM® D3606 calibration standard #1 with sec-butanol internal standard (3 components)
Catalog # REST30661
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    ASTM D3606 calibration standard #1 with sec-butanol internal standard
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
Consists of three components, those are benzene, 2-butanol and toluene.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.