Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Benzo(ghi)perylene

Benzo(ghi)perylene, 1000 µg/ml in methylene chloride, 1 ml/ampoule, 1 mL
  REST31273
 :   Restek
Benzo(ghi)perylene
  REST31273
 :  Restek
 :  
Benzo(ghi)perylene
Benzo(ghi)perylene

Benzo(ghi)perylene, 1000 µg/ml in methylene chloride, 1 ml/ampoule, 1 mL

 :  
 
£78.10
   
1 * 1 mL
 

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

 

 

  • Concentration:
    1000 µg/ml in methylene chloride
  • Pk:
    1 mL
  • Description:
    Benzo(ghi)perylene
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST31273
  • Formula:
    C₂₂H₁₂
  • Description:
    Benzo(ghi)perylene
  • CAS No.:
    191-24-2

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.