Organic reference standard, nonylphenol diethoxylate-branched isomer, CIL
Catalog # PROOCILULM71471.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1,2 mL
- Standards:Reference standard
- Description:Nonylphenol diethoxylate-branched isomer, 100 µg/ml
- Formula:C₁₉H₃₂O₃
- Description:Nonylphenol diethoxylate-branched isomer