Freezing Boxes
Supplier: DISTRIBUTOR
Jump to:
About this item
Freezer storage boxes for use with vials, tubes and microtubes from 0,5 to 2 ml in volume and to hold tubes up to 53,5 mm in diameter.
- Alphanumeric grid on lid for easy reference
- Stackable, with hinged lid
- Black boxes available for use with light sensitive samples
- Self adhesive grid is also available which attaches to the underneath of the box
5 Options Available Below
Product Details & Documents
Freezer storage boxes for use with vials, tubes and microtubes from 0,5 to 2 ml in volume and to hold tubes up to 53,5 mm in diameter.
- Alphanumeric grid on lid for easy reference
- Stackable, with hinged lid
- Black boxes available for use with light sensitive samples
- Self adhesive grid is also available which attaches to the underneath of the box
Product Family Options
Product Information
- ColourDimensionsAvailabilityPrice
- DIUKRTP/72101-BKFreezer storage box, 100-place
- DIUKRTP/72101-GFreezer storage box, 100-place
- DIUKRTP/72101-NFreezer storage box, 100-place
- DIUKRTP/72101-RFreezer storage box, 100-place
- DIUKRTP/72101-YFreezer storage box, 100-place