Order Entry
United Kingdom
ContactUsLinkComponent
HPLC/UHPLC columns for protein analysis, HALO® C4, 3,4 µm
HPLC/UHPLC columns for protein analysis, HALO® C4, 3,4 µm
Catalog # HISMTI93414-414
Supplier:  HICHROM
HPLC/UHPLC columns for protein analysis, HALO® C4, 3,4 µm
Catalog # HISMTI93414-414
Supplier:  HICHROM

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    3,4 µm
  • Pore size:
    400 Å
  • Max. pressure:
    600 bar
  • Length:
    50 mm
  • Seq:
    04
  • Endcapped:
    Yes
  • USP code:
    L26
  • pH range:
    2 - 9
  • Surface area:
    15 m²/g
  • Temperature limit:
    90 °C
  • Carbon load:
    0,4%
  • Column hardware:
    Stainless steel
  • Phase:
    C4
  • Pk:
    1

Specifications

About this item

HALO® HPLC and UHPLC columns are based on innovative Fused-Core particle technology. These particles are formed from a solid core and porous outer shell. HALO® columns provide high efficiencies and separation speeds but at approximately half the back pressure for the same column length. This lower pressure means that HALO® columns can be used with conventional HPLC and LC-MS systems, as well as with UHPLC systems.

  • Ultra-fast separations
  • High resolution and high efficiency
  • Lower backpressure than fully-porous HPLC columns
  • UHPLC like separation using HPLC instruments
  • Excellent ruggedness and reproducibility

The HALO® 400 Å C4 is suitable for the analysis of large biomolecules, including monoclonal antibodies and their various antibody-drug conjugates (ADCs).

Guard column holder

Product Information