Ethanol (taxed), pure for molecular biology (200 Proof), Riedel-de Haën™
Catalog # E7023-500ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- IR spectrum:complying
- Assay (GC):Min. 99.65 %
- Water (Karl Fischer):Max. 0.2 %
- Benzene (GC):Max. 1 ppm
Specification Test Results
About this item
:
:46.07 g/mol
Merck Index:12,03806
Boiling Pt:78…79 °C (1.013 hPa)
Melting Pt:–115 °C
Density:0.790…0.791 g/cm3 (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
REACH:01-2119457610-43
:GHS07
:GHS02
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568