Methyl formate 99%, Spectrophotometric Grade, Riedel-de Haën™
Catalog # 259705-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.01 %
- Absorbance at 400 nm:Max. 0.01
- Absorbance at 310 nm:Max. 0.01
- Absorbance at 270 nm:Max. 0.04
- Absorbance at 265 nm:Max. 0.20
- Absorbance at 260 nm:Max. 0.70
- Absorbance at 259 nm:Max. 1.0
- APHA:Max. 10
- Vapor Pressure:32.81 psi ( 55 °C)
- Refractive Index:n20/D 1.343 (lit.)
- Autoignition Temperature:842 °F
Specification Test Results
About this item
:
:60.05 g/mol
Merck Index:12,06153
Boiling Pt:32.3 °C
Melting Pt:-100.4 °C
Melting Pt:–100.4 °C
Density:0.970 g/cm3
Flash Pt:-19 °C
Flash Pt:–19 °C
Storage Temperature:Ambient
EINECS:203-481-7
UN:1243
ADR:3,I
:GHS07
:GHS02
CAS Number:107-31-3
Formula:C₂H₄O₂
MDL Number:MFCD00003291