


Sodium peroxodisulphate ≥99.0% (RT), Purum p.a.
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7775-27-1
:
Formula:Na₂S₂O₈
MDL Number:MFCD00003501
:238.11 g/mol
Merck Index:13,08729
Melting Pt:180 °C
EINECS:231-892-1
UN:1505
ADR:5.1,III
:GHS03
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:complying
- Assay:Min. 99.0 %
- Calcium (Ca):Max. 50 ppm
- Copper (Cu):Max. 50 ppm
- Cadmium (Cd):Max. 50 ppm
- Cobalt (Co):Max. 50 ppm
- Iron (Fe):Max. 50 ppm
- Potassium (K):Max. 100 ppm
- Nickel (Ni):Max. 50 ppm
- Lead (Pb):Max. 50 ppm
- Zinc (Zn):Max. 50 ppm
- Chloride (Cl):Max. 50 ppm
Specification Test Results
Frequently Bought Together