Poly(acrylamide-co-diallyldimethylammonium chloride) 10% (w/w) in water, Sigma-Aldrich®
Catalog # SIAL409081-1L
Supplier: Merck
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Color):Colorless
- Appearance (Form):Viscous Liquid
- Infrared spectrum:Conforms to Structure
- Residue on Evaporation:9.0 - 11.0 %
- Brookfield Viscosity (25 °C):9000 - 25000 cps
- pH (c = 1%; Water):5.00 - 8.00
Specification Test Results
About this item
Synonyms: P(AAm-co-DADMAC)
:
Storage Temperature:Ambient
CAS Number:26590-05-6
Formula:C₁₁H₂₁ClN₂O
MDL Number:MFCD00241413
Density:1.02 g/mL (25 °C)
Flash Pt:>100 °C
:M-Clarity™ quality level = MQ100