Analytical standard, Cimaterol, reference material, Honeywell Fluka™
Catalog # HONCR1351-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:54239-37-1
- Description:Cimaterol
- Melting point:164…169 °C
- Formula:C₁₂H₁₇N₃O
- Storage temperature:Ambient
- Description:Cimaterol
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results