Analytical standard, Tenuazonic acid-(acetyl-13C2) solution reference material, Honeywell Fluka™
Catalog # HONCR1323-2ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:2 mL
- Standards:Analytical standard, single component solution
- CAS No.:1486471-66-2
- Description:Tenuazonic acid-(acetyl-13C2) solution, 50 μg/ml in acetonitrile
- Melting point:–45 °C
- Formula:¹³C₂C₈H₁₅NO₃
- Storage temperature:–20°C
- Description:Tenuazonic acid-(acetyl-13C2) solution
- Boiling point:82 °C
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results