Analytical standard, Ethyl 4-hydroxybenzoate-ring-¹³C₆ solution, reference material, Honeywell Fluka™
Catalog # HONCR1268-1ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:1 mL
- Standards:Analytical standard, single component solution
- Description:Ethyl 4-hydroxybenzoate-ring-¹³C₆ solution, 50 μg/ml in acetone
- Melting point:–95 °C
- Formula:¹³C₆C₃H₁₀O₃
- Storage temperature:2…8 °C
- Description:Ethyl 4-hydroxybenzoate-ring-¹³C₆ solution
- Boiling point:55…57 °C
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results