Analytical standard, Cambendazole-d₇, reference material, Honeywell Fluka™
Catalog # HONCR1003-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1228182-48-6
- Description:Cambendazole-d₇
- Melting point:204…209 °C
- Formula:C₁₄D₇H₇N₄O₂S
- Storage temperature:Ambient
- Description:Cambendazole-d₇
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results