Order Entry
United Kingdom
ContactUsLinkComponent
Silicone, high vacuum grease
  EDWAH024-00-036
Silicone, high vacuum grease
  EDWAH024-00-036
  • Description:
    High vacuum silicone grease, tube, 50g
  • Pk:
    1
A high vacuum grade grease, which is for use at system pressures lower than 10⁻⁶ mbar.