Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sodium hydroxide solution 0.1 M

Sodium hydroxide solution 0.1 M

2,5 L
Catalog # DIUKSO5684
Supplier:   DISTRIBUTOR
Sodium hydroxide solution 0.1 M
Catalog # DIUKSO5684
Supplier:  DISTRIBUTOR
CAS Number:  1310-73-2
Sodium hydroxide solution 0.1 M

2,5 L

CAS Number:  1310-73-2
Price
£20.20
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
Specifications

Specifications

  • Pk:
    2,5 L

Specifications