Betaine solution 5 M, Ultrapure for molecular biology
Catalog # J77507.AB
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- DNase (endo):Passes test
- DNase (exo):Passes test
- RNase:Passes test
Specification Test Results
About this item
:
:117.15 g/mol
Storage Temperature:Freezer
EINECS:203-490-6
CAS Number:107-43-7
Formula:C₅H₁₁NO₂
MDL Number:MFCD00012123