D-(+)-Glucose, anhydrous
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
Merck Index:14,04459
Melting Pt:147…153 °C
Density:1.54 g/cm³ (20 °C)
:180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
Specification Test Results
- Identity:By IR
- Specific Rotation (25°C):+52.6 to 53.2° (dry basis, c=10 in 0.012 N NH₄OH)
- Moisture (KF):≤1.0%
Specification Test Results
Frequently Bought Together