Order Entry
United Kingdom
ContactUsLinkComponent
Potassium ionophore I - cocktail B, Selectophore™ ion sensor material, Supelco®
Toxic
Potassium ionophore I - cocktail B, Selectophore™ ion sensor material, Supelco®
Catalog # 99373-0.1ML-F
Supplier:  Merck
CAS Number:  
undefined
Toxic
Potassium ionophore I - cocktail B, Selectophore™ ion sensor material, Supelco®
Catalog # 99373-0.1ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    0,1 mL
  • Seq:
    2010

Specifications

About this item

 :
UN:3172
ADR:6.1,II
 :GHS06