Water, TECHNICAL, demineralised
23597.980
:
- Pk:1.000 L
- Seq:5037
- Conductivity (20°C) (at production):Max. 1.5 µS/cm
:
:18.02 g/mol
:13,10098
:100 °C (1013 hPa)
:0 °C
:1 g/cm³ (4 °C)
:Ambient
:231-791-2
:7732-18-5
:H₂O
:MFCD00011332