Order Entry
United Kingdom
ContactUsLinkComponent
Connectors, 1/16" male
  18-1147-10
 :  Cytiva
Connectors, 1/16" male
  18-1147-10
 :  Cytiva
  • Description:
    Fingertight narrow head connector, O. D. 1,58 mm
  • Material:
    PEEK
  • Pk:
    1
Made of Polyetheretherketone (PEEK).
For 1/16" O. D. tubing, 5/16" male.