Streptavidin, HRP (horseradish peroxidase)
Catalog # PIERN100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Conjugation:HRP (Horseradish Peroxidase)
- Storage conditions:Store at 2-8˚C upon receipt. Protect from exposure to light. Stable until expiration date if stored under these conditions.
- Protein/peptide name:Streptavidin
- Concentration:1,25 mg/ml
Specifications
About this item
HRP-conjugated streptavidin consists of streptavidin protein that is covalently conjugated to horseradish peroxidase (HRP) enzyme.
Streptavidin binds to biotin and the conjugated HRP provides enzyme activity for detection using an appropriate substrate system and also used primarily in sandwich ELISA applications to provide consistent measurement of biotinylated detection antibodies.
Caution: For research use only. Not for use in diagnostic procedures.