Tri(propylene glycol)butyl ether mixture of isomers ≥95%, Sigma-Aldrich®
SIAL438871-1L
: Merck
:
- Pack type:Poly bottle
- Pk:1 L
- Seq:5037
- Appearance (Color):Colorless
- Appearance (Form):Liquid
- Infrared spectrum:Conforms to Structure
- GC Purity (sum of isomers):≥ 94.5 %A
:
:248.36 g/mol
:276 °C (lit.)
:–75 °C (lit.)
:0.932 g/mL (25 °C) (lit.)
:M-Clarity™ quality level = MQ200
:Ambient
:55934-93-5
:C₁₃H₂₈O₄
:MFCD00216640