Order Entry
United Kingdom
ContactUsLinkComponent
2-Amino-6-methylpyridine 98%
Toxic
2-Amino-6-methylpyridine 98%
Certificates
undefined
Toxic
2-Amino-6-methylpyridine 98%

Specification Test Results

  • Appearance:
    Colorless to white to yellow to light orange Low melting crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    40 to 45 °C
  • GC:
    ≥97.5 %
  • Particle size:
    ≤0.4 % (dust)
  • pH:
    3.5 to 6

Specification Test Results

Related Information

Frequently Bought Together