
Isonicotinic acid
Catalog # MOLE63642461-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:123.11 g/mol
Merck Index:13,05206
Boiling Pt:396 °C (1013 hPa)
Melting Pt:315…319 °C
Storage Temperature:Ambient
EINECS:200-228-2
:GHS07
CAS Number:55-22-1
Formula:C₆H₅NO₂
MDL Number:MFCD00006429