
Nicotinic acid (Niacin)
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:59-67-6
:
Formula:C₆H₅NO₂
MDL Number:MFCD00006391
:123.11 g/mol
Merck Index:13,06552
Boiling Pt:258 °C (1013 hPa)
Melting Pt:142 °C
Density:1.473 g/cm³ (20 °C)
Flash Pt:193 °C
EINECS:200-441-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together