Ethyl isonicotinate, Sigma-Aldrich®
Catalog # SIAL104736-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:151.16 g/mol
Merck Index:13,05206
Boiling Pt:220 °C (1013 hPa)
Density:1.1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:216-379-2
CAS Number:1570-45-2
Formula:C₈H₉NO₂
MDL Number:MFCD00006428