Methyl nicotinate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:93-60-7
:
Formula:C₇H₇NO₂
MDL Number:MFCD00006388
:137.14 g/mol
Merck Index:12,06174
Boiling Pt:209 °C (1013 hPa)
Melting Pt:38…43 °C
Density:1,0048 g/cm³ (20 °C)
Flash Pt:95 °C
EINECS:202-261-8
:GHS07
Specification Test Results
- Appearance:White to brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:38 to 43 °C
- Titration with HClO4:≥98.5 %
Specification Test Results
Frequently Bought Together