Order Entry
United Kingdom
ContactUsLinkComponent
Isoniazid for synthesis, Sigma-Aldrich®
Isoniazid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Isoniazid for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    125 g/l (20 °C)
  • Melting Point:
    169 - 172 °C
  • Molar Mass:
    137.14 g/mol
  • Bulk Density:
    800 kg/m3
  • Flash Point:
    250 °C
  • pH:
    6 - 8 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together