2-Bromo-3-hydroxypyridine
Catalog # 8.41621.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-Bromo-3-pyridinol
:
:174 g/mol
EINECS:229-547-5
:GHS07
Storage Temperature:Ambient
CAS Number:6602-32-0
Formula:C₅H₄BrNO
MDL Number:MFCD00006220
Boiling Pt:232
Melting Pt:185…188 °C