β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Cozymase, beta-DPN, Coenzyme I, beta-Nicotinamide Adenine dinucleotide hydrate, oxidized form, beta-Diphosphopyridine Nucleotide, beta-NAD
CAS Number:53-84-9
:
Formula:C₂₁H₂₇N₇O₁₄P₂·H₂O
MDL Number:MFCD00036253
Merck Index:13,06370
Melting Pt:140 to 142 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:200-184-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together