Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
3-Methylxanthine

3-Methylxanthine

Glass bottle for solids, 100 mg
Catalog # PROOMM0176.03
Irritant
3-Methylxanthine
Catalog # PROOMM0176.03
CAS Number:  1076-22-8
3-Methylxanthine

Glass bottle for solids, 100 mg

CAS Number:  1076-22-8
Price
£1,530.00
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  3,7-Dihydro-3-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3-methylpurine
  • CAS Number:1076-22-8
  • Formula:C₆H₆N₄O₂
  • MDL Number:MFCD00005580
  • Molecular Weight:166.14 g/mol
  • EINECS:214-058-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications