3-Methylxanthine
Catalog # MOLEM28499276
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:166.14 g/mol
EINECS:214-058-1
:GHS07
CAS Number:1076-22-8
Formula:C₆H₆N₄O₂
MDL Number:MFCD00005580