Hypoxanthine, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: 1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
- CAS Number:68-94-0
- Formula:C₅H₄N₄O
- MDL Number:MFCD00005725
- Molecular Weight:136.11 g/mol
- Merck Index:13,04895
- Melting Pt:> 310 °C
- :M-Clarity™ quality level = MQ200
- EINECS:200-697-3
3 Options Available Below
Product Details & Documents
Synonyms: 1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
- CAS Number:68-94-0
- Formula:C₅H₄N₄O
- MDL Number:MFCD00005725
- Molecular Weight:136.11 g/mol
- Merck Index:13,04895
- Melting Pt:> 310 °C
- :M-Clarity™ quality level = MQ200
- EINECS:200-697-3



