Order Entry
United Kingdom
ContactUsLinkComponent
BCIP/NBT, liquid substrate ready-to-use, Sigma-Aldrich®
BCIP/NBT, liquid substrate ready-to-use, Sigma-Aldrich®
Catalog # SIALB1911-100ML
Supplier:  Merck
CAS Number:  
BCIP/NBT, liquid substrate ready-to-use, Sigma-Aldrich®
Catalog # SIALB1911-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance (Turbidity):
    Clear
  • Appearance (Color):
    Faint Green-Yellow to Light Green-Yellow to Light Yellow
  • Appearance (Form):
    Liquid
  • pH:
    8.5 - 9.5
  • BCIP Immunoblot Test:
    Compares (Compares to Powdered Substrate Mixture)

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator