Order Entry
United Kingdom
ContactUsLinkComponent
Sodium dehydracetate for synthesis, Sigma-Aldrich®
Irritant
Sodium dehydracetate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Sodium dehydracetate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    330 g/l (20 °C)
  • Melting Point:
    280 - 285 °C
  • Molar Mass:
    190.13 g/mol
  • Bulk Density:
    500 - 550 kg/m3
  • pH:
    8.8 - 9.3 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    360 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together